Anti-PAK2 Rabbit Polyclonal Antibody
Catalog # 77044-130
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:C-t-PAK2
- Antigen symbol:PAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13177
- Antigen synonyms:hPAK65|p21 activated kinase 2|EC 2.7.11.1|Gamma-PAK|CB422|Gamma PAK|Kinase|p21 (CDKN1A) activated kinase 2|p21 (CDKN1A)-activated kinase 2a
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PAK2 around the phosphorylation site of Ser197 (amino acids 163-212)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77044-130
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PAK2 (phospho Ser197) for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:1000
Type: Primary
Antigen: PAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat