Anti-CKII alpha Rabbit Polyclonal Antibody
77043-438
- Antibody type:Primary
- Antigen name:Casein kinase 2 alpha 1 polypeptide
- Antigen symbol:CKII alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P68400
- Antigen synonyms:CK2 catalytic subunit alpha|Casein kinase II alpha 1|Casein kinase II alpha 1 subunit|Casein kinase II alpha subunit|CK II alpha|CK2 alpha|Casein kinase II subunit alpha|CK II|CK2A1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Casein Kinase II alpha around the phosphorylation site of Tyr255 (amino acids 221-270)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77043-438
- Packaging:Plastic vial
Rabbit polyclonal antibody to Casein Kinase II alpha (phospho Tyr255) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: CKII alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat