Anti-Cyclin C Rabbit Polyclonal Antibody
77043-070
- Antibody type:Primary
- Antigen name:ccnc
- Antigen symbol:Cyclin C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P24863
- Antigen synonyms:CycC|Cyclin C|CCNC_HUMAN|hSRB11|Cyclin-C|OTTHUMP00000016897|SRB11 homolog
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Cyclin C around the phosphorylation site of Ser275 (amino acids 234-283)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. no.:77043-070
- Packaging:Plastic vial
Rabbit polyclonal antibody to Cyclin C (phospho Ser275) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: Cyclin C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat