Anti-CD222 Mouse Monoclonal Antibody [Clone: MEM-238] (APC (Allophycocyanin))
76953-120
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:300 kDa mannose 6 phosphate receptor
- Antigen symbol:CD222
- Clonality:Monoclonal
- Clone:MEM-238
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Size:100 T
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P11717
- Antigen synonyms:CI MPR|300 kDa mannose 6-phosphate receptor|CI Man-6-P receptor|Cation-independent mannose-6-phosphate receptor|CI-M6PR|Cation independent mannose 6 phosphate receptor|CD222 antigen|CD222|CI Man 6 P receptor
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Vaccinia virus encoding CD222
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. no.:76953-120
- Packaging:Glass bottle
Mouse monoclonal [MEM-238] antibody to CD222 (APC) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD222
Clonality: Monoclonal
Clone: MEM-238
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates