Anti-Smad2 (phospho Thr220) Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Smad2 (phospho Thr220) for IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC
- Recommended Dilutions: IHC: 1:50-1:200
Type: Primary
Antigen: Smad2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77203-238
- Form:
- Liquid
- Shipping temperature:
- Shipped on blue ice at 4 °C
Frequently Bought Together