Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:CD 162
- Antigen symbol:CD162
- Clonality:Recombinant
- Clone:rPSGL1/1601
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q14242
- Antigen synonyms:CD162 antigen|P selectin glycoprotein ligand 1|Cutaneous lymphocyte associated associated antigen|CLA|P-selectin glycoprotein ligand 1|CD162|PSGL-1|P selectin glycoprotein ligand 1 precursor|PSGL 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:43 - 120 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human CD162 protein
- Purification:Protein A/G chromatography
- Cat. no.:76961-284
- Packaging:Plastic vial
Recombinant mouse monoclonal [rPSGL1/1601] antibody to CD162 for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: CD162
Clonality: Recombinant
Clone: rPSGL1/1601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human