Anti-NKX3.1 Mouse Monoclonal Antibody [clone: NKX3.1/2836]
Catalog # 76968-746
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:BAPX 2
- Antigen symbol:NKX3.1
- Clonality:Monoclonal
- Clone:NKX3.1/2836
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q99801
- Antigen synonyms:NK homeobox (Drosophila) family 3 A|BAPX2|Homeobox protein Nkx3.1|NK homeobox, family 3, member A|NK3 homeobox 1|NK homeobox family 3 A|Homeobox protein Nkx 3.1|Homeobox protein Nkx-3.1|Homeobox protein NK-3 homolog A
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 92-224, of human NKX3.1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76968-746
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [NKX3.1/2836] antibody to NKX3.1 for ELISA and IHC-P with samples derived from Human.
- Validated Applications: ELISA, IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: NKX3.1
Clonality: Monoclonal
Clone: NKX3.1/2836
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human