Anti-OR6B2 Rabbit Polyclonal Antibody
77069-692
- Antibody type:Primary
- Antigen name:Olfactory receptor 6B2
- Antigen symbol:OR6B2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6IFH4
- Antigen synonyms:Olfactory receptor OR2-1|Olfactory receptor family 6 subfamily B member 2|OR6B2|OR6B2_HUMAN|Olfactory receptor OR2 1|OR6B2P
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human OR6B2 (amino acids 261-310)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77069-692
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR6B2 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: OR6B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human