Anti-ZNF498 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77073-144
Supplier: Antibodies.com
Supplier #: A100258-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to ZNF498 for WB and ELISA with samples derived from Human.
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:4000
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: ZNF498
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide.
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Molecular weight:35 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:Zinc finger and SCAN domain-containing protein 25
- Cat. No.:77073-144
- Reactivity:Human
- Gene ID:UniprotID# Q6NSZ9
- Antigen synonyms:Zinc finger and SCAN domain-containing protein 25|Zinc finger protein 498|ZSCAN25
- Immunogen:Synthetic peptide derived from human ZNF498 (amino acids 331-380)
- Conjugation:Unconjugated
- Antigen symbol:ZNF498
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



