Anti-Fyn Rabbit Polyclonal Antibody
Catalog # 77079-202
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:C syn protooncogene
- Antigen symbol:Fyn
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P06241
- Antigen synonyms:FYN_HUMAN|Protein tyrosine kinase fyn|p59-Fyn|Proto-oncogene c-Fyn|Fyn|FYN oncogene related to SRC FGR YES|OKT3 induced calcium influx regulator|Proto oncogene tyrosine protein kinase fyn|P59 FYN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:60 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human Fyn (amino acids 488-537)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77079-202
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Fyn for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: Fyn
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse