Anti-ANO9 Rabbit Polyclonal Antibody
Catalog # 77075-204
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:ANO9
- Antigen symbol:ANO9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# A1A5B4
- Antigen synonyms:TMEM16J|PIG5|Tumor protein p53-inducible protein 5|TP53I5|Anoctamin-9|ANO9_HUMAN|p53-induced gene 5 protein|Transmembrane protein 16J
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TM16J (amino acids 481-530)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77075-204
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TM16J for IHC, IF and ELISA with samples derived from Human.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: ANO9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human