Anti-OR52N1 Rabbit Polyclonal Antibody
Catalog # 77073-658
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:O52N1_HUMAN
- Antigen symbol:OR52N1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NH53
- Antigen synonyms:Olfactory receptor 52N1|OR52N1|Olfactory receptor, family 52, subfamily N, member 1 [|Olfactory receptor OR11-61
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR52N1 (amino acids 269-318)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77073-658
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to OR52N1 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: OR52N1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human