Anti-LKB1 Rabbit Polyclonal Antibody
77073-802
- Antibody type:Primary
- Antigen name:hLKB1
- Antigen symbol:LKB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15831
- Antigen synonyms:Serine/Threonine Kinase 11|Serine/threonine protein kinase LKB1|Polarization related protein LKB1|PJS|Serine/threonine protein kinase 11|Renal carcinoma antigen NY-REN-19|Serine/threonine-protein kinase 11|Serine/threonine protein kinase STK11|Liver kinase B1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:48 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human LKB1 (amino acids 300-349)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77073-802
- Packaging:Plastic vial
Rabbit polyclonal antibody to LKB1 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: LKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse