Anti-RB associated KRAB repressor Rabbit Polyclonal Antibody
77078-262
- Antibody type:Primary
- Antigen name:hRBaK
- Antigen symbol:RB associated KRAB repressor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NYW8
- Antigen synonyms:RBAK_HUMAN|ZNF769|RB associated KRAB zinc finger|Zinc finger protein 769|ZNF 769|RB associated KRAB repressor|Hypothetical protein RBAK|RB-associated KRAB zinc finger protein|RB-associated KRAB repressor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:82 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RBAK (amino acids 1-50)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77078-262
- Packaging:Plastic vial
Rabbit polyclonal antibody to RBAK for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: RB associated KRAB repressor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse