Anti-IL15RA Rabbit Polyclonal Antibody
77064-250
- Antibody type:Primary
- Antigen name:AA690181
- Antigen symbol:IL15RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13261
- Antigen synonyms:CD215|IL-15RA|IL-15R-alpha|IL-15 receptor subunit alpha|Interleukin 15 receptor alpha|Interleukin 15 receptor subunit alpha|IL15RA|IL 15R alpha|I15RA_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:28 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human IL15RA (amino acids 80-129)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77064-250
- Packaging:Plastic vial
Rabbit polyclonal antibody to IL15RA for WB and ELISA with samples derived from Human and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: IL15RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat