Anti-TSEN54 Rabbit Polyclonal Antibody
Catalog # 77068-304
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:HsSen54
- Antigen symbol:TSEN54
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z6J9
- Antigen synonyms:TSEN54 tRNA splicing endonuclease subunit|SEN54_HUMAN|SEN54L|SEN54 homolog|tRNA-splicing endonuclease subunit SEN54|PCH4|TSEN54|PCH2A|tRNA-intron endonuclease SEN54
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TSEN54 (amino acids 261-310)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77068-304
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TSEN54 for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: TSEN54
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human