Anti-alpha Glucosidase II Rabbit Polyclonal Antibody
77069-564
- Antibody type:Primary
- Antigen name:alpha Glucosidase II
- Antigen symbol:alpha Glucosidase II
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q14697
- Antigen synonyms:Glucosidase II subunit alpha|GANAB|Alpha-glucosidase 2|Glucosidase II alpha|Glucosidase alpha neutral AB|G2AN|Alpha glucosidase II alpha subunit|GANAB_HUMAN|Glu II
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:106 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GANAB (amino acids 242-291)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77069-564
- Packaging:Plastic vial
Rabbit polyclonal antibody to GANAB for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: alpha Glucosidase II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse