Anti-ITPK1 Rabbit Polyclonal Antibody
Catalog # 77063-374
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:3, 4)P(3) 5/6-kinase
- Antigen symbol:ITPK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13572
- Antigen synonyms:Inositol triphosphate 5/6 kinase|4-trisphosphate 5/6-kinase|Inositol tetrakisphosphate 1 kinase|Inositol-triphosphate 5/6-kinase|Inositol 1 3 4 triphosphate 5/6 kinase|Inositol 1 3 4 trisphosphate 5/6 kinase|Inositol-tetrakisphosphate 1-kinase|Inositol 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ITPK1 (amino acids 81-130)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77063-374
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ITPK1 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: ITPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse