Anti-PPFIBP1 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77069-946
Supplier: Antibodies.com
Supplier #: A99055-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to LIPB1 for WB and ELISA with samples derived from Human and Mouse.
- Recommended dilutions: WB: 1:500 to 1:1000 ELISA: 1:1000
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: PPFIBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Molecular weight:114 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:hSGT2
- Cat. No.:77069-946
- Reactivity:Human|Mouse
- Gene ID:UniprotID# Q86W92
- Antigen synonyms:Myocyte enhancer factor 2C|Myocyte-specific enhancer factor 2C
- Immunogen:Synthetic peptide derived from human LIPB1 (amino acids 282 - 331)
- Conjugation:Unconjugated
- Antigen symbol:PPFIBP1
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7.4 with 150 mM NaCl, 0.02% sodium azide, and 50% glycerol
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



