Anti-Rad51 Rabbit Polyclonal Antibody
Catalog # 77075-060
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:BRCA1/BRCA2 containing complex, subunit 5
- Antigen symbol:Rad51
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q06609
- Antigen synonyms:hRAD51|DNA repair protein RAD51 homolog 1|DNA repair protein rhp51|HsRAD51|HsT16930|MRMV2|FANCR|BRCC 5|BRCC5
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:36 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RAD51 (amino acids 281-330)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77075-060
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to RAD51 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: Rad51
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse