Anti-RBM26 Rabbit Polyclonal Antibody
77075-114
- Antibody type:Primary
- Antigen name:1700009P03Rik
- Antigen symbol:RBM26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q5T8P6
- Antigen synonyms:acidic rich RS domain containing 2|C13orf10|cutaneous T-cell lymphoma tumor antigen se70-2|FLJ20957|C230097K14Rik|AI447318|CTCL tumor antigen se70 2|C86225|ARRS2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:113 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RBM26 (amino acids 711-760)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77075-114
- Packaging:Plastic vial
Rabbit polyclonal antibody to RBM26 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: RBM26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse