Anti-SNF5 Mouse Monoclonal Antibody [clone: ABT-INI1]
Mouse monoclonal (ABT-INI1) antibody to INI 1 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
Type: Primary
Antigen: SNF5
Clonality: Monoclonal
Clone: ABT-INI1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77073-618
- 77063-674
- Antigen symbol:
- SNF5
- SNF5
- Antigen name:
- BAF47
- BAF47
- Antigen synonyms:
- hSNF5|BRG1-associated factor 47|RTPS1|RDT|Sfh1p|INI1|Integrase interactor 1 protein|Malignant rhabdoid tumor suppressor|SMARCB1
- hSNF5|BRG1-associated factor 47|RTPS1|RDT|Sfh1p|INI1|Integrase interactor 1 protein|Malignant rhabdoid tumor suppressor|SMARCB1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-INI1
- ABT-INI1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# Q12824
- UniprotID# Q12824
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human INI-1
- Synthetic peptide derived from human INI-1
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
- IHC
Frequently Bought Together