Anti-KAT2A/GCN5 Rabbit Polyclonal Antibody
77077-658
- Antibody type:Primary
- Antigen name:1110051E14Rik
- Antigen symbol:KAT2A/GCN5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q92830
- Antigen synonyms:GCN5 general control of amino-acid synthesis 5-like 2 (yeast)|EC 2.3.1.48|general control of amino acid synthesis 5-like 2|GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2|AW212720|General control of amino acid synthesis protein 5-like 2|Gcn5|GCN5L2|GCN 5
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:93 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GCN5L2 (amino acids 691-740)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77077-658
- Packaging:Plastic vial
Rabbit polyclonal antibody to GCN5L2 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: KAT2A/GCN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse