Anti-GPCR GPR124 Rabbit Polyclonal Antibody
Catalog # 77071-162
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DKFZp434C211
- Antigen symbol:GPCR GPR124
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q96PE1
- Antigen synonyms:G-protein coupled receptor 124|KIAA1531|GP124_HUMAN|GPCR GPR124|G protein coupled receptor 124|GPR 124|DKFZp434J0911|TEM 5|FLJ14390
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:141 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GPR124 (amino acids 1151-1200)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77071-162
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GPR124 for IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: GPCR GPR124
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse