Anti-PRDM12 Rabbit Polyclonal Antibody
Catalog # 77071-544
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Gm998
- Antigen symbol:PRDM12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H4Q4
- Antigen synonyms:PR domain containing protein 12|PRD12_HUMAN|PFM9|PR domain-containing protein 12|PFM 9|PRDM12|PR domain containing 12|PRDM 12|PR domain zinc finger protein 12
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PRDM12 (amino acids 191-240)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77071-544
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PRDM12 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: PRDM12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse