Anti-GPR37L1 Rabbit Polyclonal Antibody
77072-816
- Antibody type:Primary
- Antigen name:Endothelin B Receptor Like Protein 2
- Antigen symbol:GPR37L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O60883
- Antigen synonyms:ETBR LP 2|G protein coupled receptor 37 like 1|ETBR2_HUMAN|GPR37L1|G-protein coupled receptor 37-like 1|Endothelin B receptor-like protein 2|ETBR-LP-2|ETBRLP2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ETBR2 (amino acids 1-50)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77072-816
- Packaging:Plastic vial
Rabbit polyclonal antibody to ETBR2 for WB, IHC, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: GPR37L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human