Anti-GCNT7 Rabbit Polyclonal Antibody
Catalog # 77069-476
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:3-galactosyl-O-glycosyl-glycoprotein beta-1
- Antigen symbol:GCNT7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6ZNI0
- Antigen synonyms:6-N-acetylglucosaminyltransferase 7|GCNT7|GCNT7_HUMAN|GCNT|glucosaminyl (N-acetyl) transferase family member 7|C20orf105|Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 7
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GCNT7 (amino acids 311-360)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77069-476
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GCNT7 for WB, IHC, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: GCNT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human