Anti-FOXO3A Rabbit Polyclonal Antibody
Catalog # 77073-952
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:AF6q21
- Antigen symbol:FOXO3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O43524
- Antigen synonyms:Forkhead box O3|FKHRL1P2|DKFZp781A0677|Forkhead box O3A|FKHRL 1|FKHRL1|Forkhead (Drosophila) homolog (rhabdomyosarcoma) like 1|AF6q21 protein|FKHR2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:71 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human FOXO3A (amino acids 281-330)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77073-952
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to FOXO3A for IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: FOXO3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse