Anti-Rabenosyn 5 Rabbit Polyclonal Antibody
Catalog # 77078-550
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:110 kDa protein
- Antigen symbol:Rabenosyn 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H1K0
- Antigen synonyms:RBNS5_HUMAN|zinc finger FYVE domain containing 20|Zfyve20|FYVE finger-containing Rab5 effector protein rabenosyn-5|Zinc finger FYVE domain-containing protein 20|FYVE finger containing Rab5 effector protein Rabenosyn 5
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:88 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ZFYVE20 (amino acids 145-194)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77078-550
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ZFYVE20 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Rabenosyn 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse