Anti-Lymphotoxin Beta Rabbit Polyclonal Antibody
77076-930
- Antibody type:Primary
- Antigen name:LT beta
- Antigen symbol:Lymphotoxin Beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q06643
- Antigen synonyms:TNF-C|Lymphotoxin-beta|Lymphotoxin Beta|p33|TNFSF3|TNFC_HUMAN|TNFC|LT-beta|Ltb
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TNFC (amino acids 181-230)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77076-930
- Packaging:Plastic vial
Rabbit polyclonal antibody to TNFC for WB, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: Lymphotoxin Beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse