Anti-RSK4 Rabbit Polyclonal Antibody
Catalog # 77081-076
Supplier: Antibodies.com
Specifications
- Antibody type:Primary
- Antigen name:90 kDa ribosomal protein S6 kinase 6
- Antigen symbol:RSK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UK32
- Antigen synonyms:Ribosomal protein S6 kinase alpha-6|Ribosomal protein S6 kinase alpha 6|p90RSK6|p90 RSK 6|pp90RSK4|Ribosomal protein S6 kinase 90kDa polypeptide 6|KS6A6_HUMAN|p90-RSK 6|Ribosomal S6 kinase 4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:83 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human S6K-alpha6 (amino acids 661-710)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77081-076
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to S6K-alpha6 for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: RSK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human