Anti-Cadherin 9 Rabbit Polyclonal Antibody
Catalog # 77062-220
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:CADH9_HUMAN
- Antigen symbol:Cadherin 9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9ULB4
- Antigen synonyms:T1 cadherin|Cadherin9|CDH9|CDH 9|Cadherin 9|Cadherin-9|Cadherin 9 type 2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:88 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CDH9 (amino acids 173-222)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77062-220
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CDH9 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Cadherin 9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse