Anti-FKRP Rabbit Polyclonal Antibody
Catalogue # 77061-156
Fournisseur: ANTIBODIES.COM LLC
Spécifications
- Antibody type:Primary
- Antigen name:FKRP
- Antigen symbol:FKRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H9S5
- Antigen synonyms:MGC2991|Fukutin-related protein|FKRP_HUMAN|LGMD2I|Fukutin related protein|FLJ12576|MDC1C
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:54 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human FKRP (amino acids 1-50)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77061-156
- Packaging:Plastic vial
Spécifications
À propos de cet article
Rabbit polyclonal antibody to FKRP for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: FKRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat