Anti-Frizzled 4 Rabbit Polyclonal Antibody
Catalog # 77061-132
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:CD 344
- Antigen symbol:Frizzled 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9ULV1
- Antigen synonyms:CD344 antigen|CD 344 antigen|Frizzled (Drosophila) homolog 4|FEVR|Frizzled family receptor 4|Frizzled 4|CD344|EVR1|Frizzled homolog 4 (Drosophila)
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human FZD4 (amino acids 131-180)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77061-132
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to FZD4 for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: Frizzled 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat