Anti-Connexin 37/GJA4 Rabbit Polyclonal Antibody
Catalog # 77061-072
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Connexin 37
- Antigen symbol:Connexin 37/GJA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35212
- Antigen synonyms:Cx37|Gap junction protein, alpha 4, 37kDa|Connexin 37 / GJA4|Gap junction alpha 4 protein|gap junction protein, alpha 4 37kDa (connexin 37)|gap junction protein, alpha 4, 37kD (connexin 37)|CXA4_HUMAN|Gap junction alpha-4 protein|Connexin-37
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GJA4 (amino acids 81-130)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77061-072
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GJA4 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: Connexin 37/GJA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat