Anti-NRF1 Rabbit Polyclonal Antibody
Catalog # 77060-558
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:alpha pal
- Antigen symbol:NRF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q16656
- Antigen synonyms:NRF1_HUMAN|Alpha-pal|locus control region factor 1|NRF-1|NRF1|Alpha palindromic-binding protein|NFE2 related factor 1|Nuclear respiratory factor 1|alpha palindromic binding protein
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NRF1 (amino acids 221-270)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77060-558
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NRF1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: NRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat