Anti-TAK1 Rabbit Polyclonal Antibody
Catalog # 77060-130
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:M3K7_HUMAN
- Antigen symbol:TAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O43318
- Antigen synonyms:MAP3K 7|Mitogen activated protein kinase kinase kinase 7|TAK1|TGF-beta-activated kinase 1|MEKK7|Mitogen-activated protein kinase kinase kinase 7|TGF1a|TGF beta activated kinase 1|Map3k7
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TAK1 (amino acids 161-210)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77060-130
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TAK1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: TAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat