Order Entry
Canada
ContactUsLinkComponent
Anti-beta 3 Adrenergic Receptor Rabbit Polyclonal Antibody
Anti-beta 3 Adrenergic Receptor Rabbit Polyclonal Antibody
Catalog # 77059-478
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-beta 3 Adrenergic Receptor Rabbit Polyclonal Antibody
Catalog # 77059-478
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A97731-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADR B3
  • Antigen symbol:
    beta 3 Adrenergic Receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13945
  • Antigen synonyms:
    ADRB3R|ADRB3|ADRB 3|Beta 3 adrenoceptor|Adrenergic beta 3 receptor|ADRB3_HUMAN|beta 3 Adrenergic Receptor|Beta 3 adrenoreceptor|B3AR
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human ADRB3 (amino acids 250-299)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. no.:
    77059-478
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to ADRB3 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000

Type: Primary
Antigen: beta 3 Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat