Anti-CRSP2/MED14 Rabbit Polyclonal Antibody
Catalog # 77058-198
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Activator-recruited cofactor 150 kDa component
- Antigen symbol:CRSP2/MED14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O60244
- Antigen synonyms:DRIP150|MED14_HUMAN|hRGR1|CRSP complex subunit 2|EXLM1|Mediator complex subunit 14|ARC150|Cofactor required for Sp1 transcriptional activation subunit 2|CRSP2 / MED14
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:160 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human MED14 (amino acids 701-750)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77058-198
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to MED14 for IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: CRSP2/MED14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse