Anti-TRPC3 Rabbit Polyclonal Antibody
Catalog # 77057-100
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Htrp 3
- Antigen symbol:TRPC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13507
- Antigen synonyms:Mtrp3|MGC124333|Htrp3|Transient receptor protein 3|transient receptor potential cation channel subfamily C member 3 variant c|TRANSIENT RECEPTOR POTENTIAL DROSOPHILA HOMOLOG OF 3|Short transient receptor potential channel 3|Transient receptor potential cation channel subfamily C member 3|Receptor activated cation channel TRP3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:96 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human TRPC3 (amino acids 411-460)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. no.:77057-100
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TRPC3 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: TRPC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat