Anti-Uroplakin III Mouse Monoclonal Antibody [clone: ABT-UPK3]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (ABT-UPK3) antibody to Uroplakin III for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: Uroplakin III
Clonality: Monoclonal
Clone: ABT-UPK3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77056-822
- 77056-824
- Antigen symbol:
- Uroplakin III
- Uroplakin III
- Antigen name:
- MGC119178
- MGC119178
- Antigen synonyms:
- UPK 3A|UPIIIA|UPK 3|UPK3|UPK3A|UP III|OTTHUMP00000028951|UPIII|UP3a
- UPK 3A|UPIIIA|UPK 3|UPK3|UPK3A|UP III|OTTHUMP00000028951|UPIII|UP3a
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-UPK3
- ABT-UPK3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# O75631
- UniprotID# O75631
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Uroplakin III
- Synthetic peptide derived from human Uroplakin III
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
- IHC
Specifications
Related Information
Frequently Bought Together