Anti-alpha 1a Adrenergic Receptor Rabbit Polyclonal Antibody
77056-438
- Antibody type:Primary
- Antigen name:ADA1A_HUMAN
- Antigen symbol:alpha 1a Adrenergic Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35348
- Antigen synonyms:Adrenergic, alpha 1A , receptor variant 11|ADRA1L1|ADRA1A|Adrenergic, alpha 1A , receptor variant 1|Adrenergic receptor alpha 1c|Adrenergic alpha1A receptor|Adra1c|Adrenergic receptor alpha 1a|Adrenergic alpha1C receptor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:51 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ADRA1A (amino acids 136-185)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77056-438
- Packaging:Plastic vial
Rabbit polyclonal antibody to ADRA1A for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: alpha 1a Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat