Anti-alpha 1b Adrenergic Receptor Rabbit Polyclonal Antibody
Catalog # 77056-434
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:ADA1B_HUMAN
- Antigen symbol:alpha 1b Adrenergic Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P35368
- Antigen synonyms:Adrenergic alpha 1B receptor|Alpha 1 adrenergic receptor|ADRA1B|Adrenoceptor alpha 1B|Adrenergic receptor alpha 1b|alpha 1b Adrenergic Receptor|Adra 1b|ADRA 1|ADRA1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ADRA1B (amino acids 431-480)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77056-434
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ADRA1B for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: alpha 1b Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat