Anti-GPR180 Rabbit Polyclonal Antibody
Catalog # 77056-050
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:G protein coupled receptor 180
- Antigen symbol:GPR180
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q86V85
- Antigen synonyms:Intimal thickness related receptor|GP180_HUMAN|GPR180|Itr|Intimal thickness-related receptor|GPR 180|Integral membrane protein GPR180
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GPR180 (amino acids 231-280)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77056-050
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GPR180 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: GPR180
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse