Anti-SLC9A8 Rabbit Polyclonal Antibody
Catalog # 77055-462
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Na(+)/H(+) exchanger 8
- Antigen symbol:SLC9A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y2E8
- Antigen synonyms:NHE 8|Sodium/ hydrogen exchanger 8|Sodium/hydrogen exchanger 8|SL9A8_HUMAN|Solute carrier family 9 member 8|NHE-8|solute carrier family 9 (sodium/hydrogen exchanger), member 8|SLC9A8|NHE8
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:64 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC9A8 (amino acids 440-489)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77055-462
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SLC9A8 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: SLC9A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat