Anti-EIF1AY Rabbit Polyclonal Antibody
Catalog # 77055-102
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:eIF 1A Y isoform
- Antigen symbol:EIF1AY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O14602
- Antigen synonyms:EIF1AY|eIF-1A Y isoform|IF1AY_HUMAN|Eukaryotic translation initiation factor 1A|Eukaryotic translation initiation factor 1A, Y chromosomal|Eukaryotic translation initiation factor 4C|Eukaryotic translation initiation factor 1A, Y linked|EIF-4C|eIF 4C
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:16 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human EIF1AY (amino acids 21-70)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77055-102
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to EIF1AY for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:5000
Type: Primary
Antigen: EIF1AY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat