Anti-PPP2R3C Rabbit Polyclonal Antibody
Catalog # 77054-778
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:C14orf10
- Antigen symbol:PPP2R3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q969Q6
- Antigen synonyms:protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma|PPP2R3C|protein phosphatase 2, regulatory subunit B'', gamma|Protein phosphatase 2 regulatory subunit B''gamma|P2R3C_HUMAN|Hypothetical protein FLJ20644|G5pr|Chromosome 14 open reading frame 10|G4-1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PPP2R3C (amino acids 165-214)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77054-778
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PPP2R3C for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: PPP2R3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat