Anti-SLMAP Rabbit Polyclonal Antibody
Supplier: Antibodies.com
Certificates
About this item
Rabbit polyclonal antibody to SLMAP for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: SLMAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77202-348
- Antigen symbol:
- SLMAP
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q14BN4
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 540-790 of human SLMAP (NP_009090.2)
- Sequence:
- IDTENLREEKDSEITSTRDELLSARDEILLLHQAAAKVASERDTDIASLQEELKKVRAELERWRKAASEYEKEITSLQNSFQLRCQQCEDQQREEATRLQGELEKLRKEWNALETECHSLKRENVLLSSELQRQEKELHNSQKQSLELTSDLSILQMSRKELENQVGSLKEQHLRDSADLKTLLSKAENQAKDVQKEYEKTQTVLSELKLKFEMTEQEKQSITDELKQCKNNLKLLREKGNNKPWPWMPML
- Form:
- Liquid
- Molecular weight:
- 110 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together