About this item
This VIP ECO series of compact, ultra-low temperature (−86 °C) freezers are ideal for personal or individual clinical usage. The series makes use of a natural refrigerant, reducing environmental impact and delivering optimal storage in a compact footprint.
- Energy efficient performance
- Organic EL offers multiple display options
- Door latch and magnetic gasket combined
- Compact, personal-use type installs anywhere
Natural refrigerants: Naturally occurring hydrocarbon (HC) refrigerants have minimal effect on the environment and are compliant with environmental legislation for climate control. These refrigerants also provide efficient cooling without compromising performance, ambient tolerance, and recovery speeds following door openings.
Organic EL control panel includes USB port: The control panel with organic EL display is positioned close to the top to help enhance ease of use. The enhanced visibility makes user operation simple and intuitive. A logging function is included, allowing users to save temperature/alarm history data and transfer data using the USB port.
Warranty: 5 years parts and labor.
Specifications
- Catalog No:
- CA77282-390
- CA77282-388
- Model:
- MDF-DC202VH-PA
- MDF-DC102VH-PA
- Cat. No.:
- CA77282-390
- CA77282-388
- Refrigerant:
- Mixed HC refrigerant (R-290, R-170, R-50)
- Mixed HC refrigerant (R-290, R-170, R-50)
- Compressor:
- 550 W
- 550 W
- Electrical:
- 115 V, 60 Hz
- 115 V, 60 Hz
- Noise level:
- 52 dB(A)
- 52 dB(A)
- Material:
- Painted steel
- Painted steel