Order Entry
Canada
ContactUsLinkComponent
Human TIGIT ELISA Kit
Human TIGIT ELISA Kit
Catalog # 77210-776
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human TIGIT ELISA Kit
Catalog # 77210-776
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A310257-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    TIGIT
  • Description:
    Human TIGIT ELISA kit
  • Size:
    96 tests
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0.75 - 12 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0.019 ng/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    77210-776

Specifications

About this item

Human TIGIT ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TIGIT in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 0.75 to 12 ng/mL
  • Sensitivity: 0.019 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human TIGIT ELISA kit (A310257) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TIGIT in An antibody specific for TIGIT has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TIGIT present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-TIGIT antibody, which also binds the TIGIT present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-TIGIT antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-TIGIT antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of TIGIT captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of TIGIT in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.